Miniature Valve, Inert, Ace Glass Incorporated
Supplier: Ace Glass
5850-10
89052-924EA 327.85 USD
89052-924
Miniature Valve, Inert, Ace Glass Incorporated
Valves Vacuum Valves
This miniature straight-through shut-off valve is designed for small volumes of ultrapure or corrosive fluids
Liquids touch only inert PTFE or inert CTFE. The housing features ¼"–28 UNF female threaded ports with 0.059" I.D. holes to accept a variety of fittings.