Adapter, Three Matching Ace-Thred Connections, Ace Glass Incorporated
Supplier: Ace Glass
5829-04 5829-08 5829-12 5829-14
89052-478EA 147.02 USD
89052-478 89052-480 89052-482 89052-484
Adapter, Three Matching Ace-Thred Connections, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is threaded at both ends and on side joint with matching Ace-Thred connections.
  • Top, bottom and side joints
  • Matching thread connections
  • Fabricated from borosilicate glass
Ace-Thred size listed also refers to the I.D. of the tubing above the lower thread. Fabricated from borosilicate glass.