Chromatography Column, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5821-05 5821-09 5821-112 5821-13 5821-15 5821-17 5821-26 5821-28 5821-29 5821-30 5821-31 5821-32 5821-24
89052-396EA 378.96 USD
89052-396 89052-398 89052-400 89052-402 89052-404 89052-406 89052-410 89052-412 89052-414 89052-416 89052-418 89052-420 89052-408
Chromatography Column, Jacketed, Ace Glass Incorporated
Chromatography Columns
Columns are fully jacketed, including the internally threaded ends, for circulation of heat exchange media.
A wide choice of upper and lower end pieces can be fitted to columns. Fabricated from borosilicate glass. The column int.Ø denotes Minimum Value. The Effective Length refers Approximate.