Standard Taper Joint Adapter, Ace Glass
Supplier: Ace Glass
5295-14 5295-16 5295-18 5295-22 5295-30 5295-12
89051-550EA 88.48 USD
89051-550 89051-552 89051-554 89051-556 89051-560 89082-640
Standard Taper Joint Adapter, Ace Glass
Adapters Adapters, Joints with Tube Connectors
Adapters can be ordered as is or can be modified to meet your application.
  • Can be supplied to fit flask capacities up to 72L
  • Hose connection on -12 and -14
  • Fabricated from borosilicate glass
Please specify either the length needed below the joint, or the capacity, or the Part number of the vessel into which the adapter is being placed. Will supply the bottom tube with the proper curvature, if required to fit the vessel. These can be supplied to fit flask capacities up to 72L. Hose connection on -12 and -14 is size D for 3/8" tubing.
Ordering information: Product number 89051-558 is for use as a thermometer, thermoprobe holder, or a plain vent. Tubes have three different, bottom, ground inner standard taper joint sizes.