Adapter, "Midi" #11 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5261-37 5261-35 5261-36 5261-39 5261-38 5261-40
89051-490EA 101.72 USD
89051-490 89051-494 89051-496 89051-502 89051-508 89051-512
Adapter, "Midi" #11 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "midi" adapter has a standard taper inner joint at the bottom, #11 Ace-Thred at the top and a serrated hose connection.
  • Ace-Thred at top
  • Bottom [ST] inner joint
  • Serrated hose connection
  • Fabricated from borosilicate glass
Adapter will accept inner tubes with diameters of 9mm to 10.5mm such as thermowells, gas dispersion tubes, and vacuum takeoffs. Suitable for most vacuum work.
Ordering information: Available complete with a 89059-214 nylon bushing and 89060-966 FETFE o-ring, or as glassware only.