Adapter, Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5217-11 5217-35 5217-23 9070-02 5217-10 5217-40
89051-408EA 55.38 USD
89051-408 89051-410 89051-432 89051-486 89051-506 89082-466
Adapter, Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a hose connection at one end.
  • Ground [ST] outer or spherical socket joint
  • With hose connection
  • Fabricated from borosilicate glass
This adapter has a ground standard taper outer or spherical socket joint at one end and a hose connection at the other end.