Adapter, Vacuum, Ace Glass Incorporated
Supplier: Ace Glass
5260-07 5260-10 5260-15 5260-35 9123-06 9123-08
89051-476EA 128.41 USD
89051-476 89051-478 89051-480 89051-482 89064-994 89064-998
Adapter, Vacuum, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This vacuum adapter comes with side hose connections and a drip tip.
  • Side hose connections
  • Drip tip
  • Standard taper or spherical joints
  • Fabricated from borosilicate glass
Adapter is available with either standard taper or spherical joints and is fabricated from borosilicate glass.