Adapter, "Maxi" #15 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5261-58 5261-56 5261-57 5261-59
89051-422EA 112.02 USD
89051-422 89051-428 89051-500 89051-504
Adapter, "Maxi" #15 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter has a standard taper inner joint at the bottom, #15 Ace-Thred at the top and a serrated hose connection.
  • Ace-Thred at top
  • Bottom [ST] inner joint
  • Serrated hose connection
  • Fabricated from borosilicate glass
Adapter will accept inner tubes with diameters of 12.5mm to 14mm such as electrodes, and inlet and outlet tubes. Suitable for most vacuum work.
Ordering information: Available complete with a 89059-220 nylon bushing and 89060-984 FETFE o-ring, or as glassware only.