Adapter, PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
5210-40
89051-416EA 178.5 USD
89051-416
Adapter, PTFE Stopcock, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a hose connection and PTFE stopcock plug.
  • Reinforced [ST] outer joint
  • PTFE stopcock plug
  • With hose connection
  • Fabricated from borosilicate glass
Adapter has a reinforced standard taper outer joint, a hose connection, and 1:5 solid PTFE stopcock plug. Plug bore is 2mm. For stopcock plug, see 89062-770. Use with 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.