Adapter, [SJ], Angled Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5200-35
89051-396EA 188.18 USD
89051-396
Adapter, [SJ], Angled Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a 90 degree angled hose connection with a spherical ball joint at the bottom and glass stopcock.
  • 90° angled hose connection
  • Bottom spherical ball joint
  • Glass stopcock
  • Fabricated from borosilicate glass
Adapter has a glass stopcock with a 3mm plug bore. Bore size represents the plug bore size.