Adapter, [ST], Angled Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5200-05 5200-10 5200-15 9080-02 9080-08 9080-10
89051-390EA 151.37 USD
89051-390 89051-392 89051-394 89064-848 89064-850 89064-852
Adapter, [ST], Angled Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a 90 degree angled hose connection with inner standard taper joint at bottom and glass stopcock.
  • 90° angled hose connection
  • Bottom inner [ST] joint
  • Glass stopcock
  • Fabricated from borosilicate glass
Adapter features a glass stopcock with a 2mm or 3mm bore.