Adapter, Vacuum Take-Off, Short Stem, Ace Glass Incorporated
Supplier: Ace Glass
5193-08 5193-14
89051-376EA 85.56 USD
89051-376 89082-824
Adapter, Vacuum Take-Off, Short Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This vacuum take-off adapter features a short stem.
  • Stem length below joint of 30 mm
  • With hose connection
  • Fabricated from borosilicate glass
The stem length below the joint is 30mm. Use with a 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.