Adapter, Short Stem, Ace Glass Incorporated
Supplier: Ace Glass
5192-45
89051-368EA 131.73 USD
89051-368
Adapter, Short Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The hose connection on the side of this adapter faces the same direction as top joint.
  • 105° angle outer joint
  • Side hose connection
  • Fabricated from borosilicate glass
Featuring an outer joint that is at an angle of 105°. The hose connection on the side faces the same direction as top joint. Stem below the joint is 30mm. Use with a 3/8-inch or 5/16-inch I.D. tubing, size C hose connection.