Adapter, Liquid Inlet, Ace Glass Incorporated
Supplier: Ace Glass
5175-10
89051-358EA 102.8 USD
89051-358
Adapter, Liquid Inlet, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter has a standard taper inner joint and two vacuum take-offs.
  • [ST] inner joint
  • Two vacuum take-offs
  • Fabricated from borosilicate glass
Featuring a standard taper 24/40 inner joint. Use 3/8-inch I.D. tubing, size D hose connection.