Adapter, Offset, Ace-Threds, Ace Glass Incorporated
Supplier: Ace Glass
5102-05
89051-324EA 113.22 USD
89051-324
Adapter, Offset, Ace-Threds, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features an Ace-Thred at the bottom and two smaller Ace-Threds at the top, with one being offset.
  • Bottom Ace-Thred
  • Two top Ace-Threds, one offset
  • Nylon bushings and o-rings not included
  • Fabricated from borosilicate glass
Adapter comes with a #15 Ace-Thred at the bottom and two #7 Ace-Threds at the top, with one being offset. Used with a 89063-384 temperature measurement apparatus for a 89058-884 hydrogenation/gas apparatus. Glass only, NOT supplied with 89051-128 nylon bushings or o-rings.