Adapter, Offset, Ace Glass Incorporated
Supplier: Ace Glass
5101-54
89051-322EA 142.89 USD
89051-322
Adapter, Offset, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features an Ace-Thred at the top, a reinforced standard taper outer joint on the side arm, and a standard taper inner joint at the bottom.
  • Top Ace-Thred
  • Side reinforced [ST] outer joint
  • Bottom [ST] inner joint
  • Nylon bushing and o-ring included
  • Fabricated from borosilicate glass
Adapter comes with a #7 Ace-Thred at the top, reinforced standard taper 24/40 outer joint on side arm, and standard taper inner joint at bottom. Supplied with a 89051-122 nylon bushing and a 89060-958 FETFE O-Ring.