Adapter, Distilling Trap, Ace Glass Incorporated
Supplier: Ace Glass
5230-10
89051-306EA 125.33 USD
89051-306
Adapter, Distilling Trap, Ace Glass Incorporated
Distillation Components Distillation Adapters
The distilling trap adapter is used to prevent carryover of reactions that have a tendency to foam.
  • Outlet tube bent at 75° angle
  • [ST] 24/40 joint
  • Fabricated from borosilicate glass
This distilling trap adapter has an outlet tube bent at a 75 degree angle.