Adapter, 75 Degree Angle, Outlet Tube, Ace Glass Incorporated
Supplier: Ace Glass
5095-10
89051-272EA 105.48 USD
89051-272
Adapter, 75 Degree Angle, Outlet Tube, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a standard taper inner joint at the bottom and side, and an outlet tube at the top.
  • Bottom and side [ST] inner joints
  • Top outlet tube
  • Fabricated from borosicilate glass
Adapter comes with a standard taper 24/40 inner joint at the bottom and side, and a 15.8mm outer diameter x 9.5 mm inner diameter outlet tube at the top.