Adapter, 105 Degree Side Arm, Borosillicate Glass, Ace Glass Incorporated
Supplier: Ace Glass
5050-86 5050-96
89051-250EA 374.21 USD
89051-250 89051-252
Adapter, 105 Degree Side Arm, Borosillicate Glass, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a 105° side arm.
  • Side reinforced [ST] outer joint
  • Bottom [ST] inner joint
  • Top Ace-Thred joint
  • Fabricated from borosilicate glass
Adapter has a reinforced standard taper outer joint on the side, a standard taper inner joint at the bottom, and an Ace-Thred joint at the top. This adapter is not supplied with bushing and o-rings, which must be ordered separately.
Ordering information: Adapter 89051-250 may be used in conjunction with Bushing with O-Ring 89059-226, or with O-Ring 89061-016. Adapter 89051-252 may be used in conjunction with Bushing with O-Ring 89059-220, or with O-Ring 89060-984.