Adapter, Ace-Thred "Twin", Ace Glass Incorporated
Supplier: Ace Glass
5031-12 5031-86 5031-10 5031-24 5031-33
89051-184EA 185.93 USD
89051-184 89051-186 89051-194 89082-814 89082-820
Adapter, Ace-Thred "Twin", Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This adapter has an inner standard taper joint at bottom and two offset Ace-Thred ports at top.
  • Bottom inner [ST] joint
  • Two offset Ace-Thred ports at top
  • Accommodates two inner tubes
  • Two nylon bushings and FETFE o-rings included
Adapter has two threaded openings that enable you to insert two inner tubes, such as a thermometer and a bleed tube, through the same joint. Supplied complete with two nylon bushings and FETFE o-rings.