Adapter, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5028-26 5028-24 5028-25 5028-28 5028-32 5028-38 5028-42 5028-30
89051-096EA 62.23 USD
89051-096 89051-100 89051-102 89051-108 89051-114 89051-116 89051-118 89051-098
Adapter, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "Mini" adapter has a ground joint at the bottom and a threaded nylon bushing with o-ring at the top that tightens into a #7 Ace-Thred joint to form a compression seal.
  • Bottom ground joint
  • Threaded nylon bushing with o-ring at top
  • Accommodates thermometer
  • Suitable for vacuum work
  • Manufactured from borosilicate glass
Adapter features a threaded nylon bushing (complete with o-ring) at top that tightens into a #7 Ace-Thred joint to form a compression seal with thermometers, bleed tubes, etc. Standard taper 10/10 size will accommodate thermometers up to 6.4mm diameters; all others will accommodate 7mm diameter. This adapter is suitable for vacuum work.