Adapter, Electrode, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5028-117 5028-119
89051-104EA 72.54 USD
89051-104 89051-110
Adapter, Electrode, "Mini" #7 Ace-Thred, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This "Mini" electrode adapter has a ground inner standard taper joint at the bottom and threaded nylon bushing with o-ring at the top that tightens into a #7 Ace-Thred joint to form an o-ring compression seal.
  • Bottom ground inner [ST] joint
  • Threaded nylon bushing with o-ring at top
  • Suitable for most vacuum work
  • Manufactured from borosilicate glass
Adapter features a threaded nylon bushing (complete with o-ring) at the top that tightens into a #7 Ace-Thred joint to form an o-ring compression seal. This adapter is suitable for most vacuum work.