SoftGrip™ 100, 300, and 1000 µl Single Channel Adjustable Volume Manual Pipettor Kit, Hamilton Company
Supplier: Hamilton
SSKIT-02
89050-808EA 1090.27 USD
89050-808
SoftGrip™ 100, 300, and 1000 µl Single Channel Adjustable Volume Manual Pipettor Kit, Hamilton Company
Pipettes Micropipettes
The Hamilton line of pipettors puts a priority on addressing the issues of hand strain, fatigue, and other injuries.
  • Features a dual indicator that improves the volume adjustment accuracy by removing parallax errors
  • Slim ejection sleeve is made from a high strength, chemically resistant polymer
  • Design allows the pipettor to reach the bottom of narrow tubes without the need for extended length tips
SoftGrip™ pipettors have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods. The SoftGrip™ tip nozzle ensures a proper fit with universal pipettor tips. Unique color coding and labeling clearly indicates the working ranges of the adjustable volume pipettors.
Hamilton products set new standards for quality, comfort, and precision. The Single Channel Adjustable Volume Manual Pipettor Kit includes 100, 300, and 1000 µl SoftGrip™ pipettors, as well as a six-position pipettor carousel.
Ordering information: For pipettor calibration labels, see 89050-826 series. For pipettor calibration key, see 89050-824 series. For standard length Hamilton pipettor tips, see 60372-204 series.