Blood Banking Medical Specimen Transporter Totes, Sonoco ThermoSafe
Supplier: Avantor
89050-562EA 564.41 USD
89050-562
Blood Banking Medical Specimen Transporter Totes, Sonoco ThermoSafe
Coolers Transport Coolers
Insulated expanded polypropylene medical transporter totes are insulated carriers ideal for on-site collection, storage and transport of temperature-sensitive specimens, medications, medical devices, and organs.
  • Qualified by ISC Labs
  • Includes operating instructions and test data
  • Side-loading clear plastic document holder
  • Durable nylon clad transporter with expanded polypropylene container
  • Stain and impact resistant
  • Qualification report included
Insulated expanded polypropylene medical transporters are for the temporary storage and transport of 16 units of both platelet (20-24˚C for up to 8 hours), and non-platelet production whole blood (1-6˚C for 8 hours and 1-10˚C for up to 24 hours) with crushed wet ice.
Ordering information: Dolly is sold separately.