Sodium acetate trihydrate ≥99%, granular USP suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Acetic acid sodium salt trihydrate
SO105-100KGB
SO105-12KG
SO105-100KG
SO105-500GM
SO105-2.5KG
SO105-50KGBL
700004-370EA
2157.25
USD
700004-370
89050-448
89089-164
89050-452
89050-454
89050-450
Sodium acetate trihydrate ≥99%, granular USP suitable for use as excipient
Sodium acetate trihydrate
Sodium Acetate, Trihydrate, Granular, USP is a moderately water soluble crystalline source of Sodium and serves as processing aid excipient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120…123 °C (1013 hPa) Melting Pt: 58 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...