VWR® Urine Sediment Processing Tube
Supplier: VWR International
89049-036
89049-036CS 409.1 USD
89049-036
VWR® Urine Sediment Processing Tube
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Disposable polystyrene tubes are designed for use with Sysmex® UF series urine analyzers.
  • Graduated markings
  • Disposable design
  • Polystyrene construction
  • For use with urine analyzers
Graduated to 10 mL, these tubes offer a 12 mL capacity that includes a 400µL collection nipple reservoir. The unique tube design features straight sides that allow for placement in each hole of testing racks. Maximum RCF: 1500g.
Ordering information: Use with cap 82028-226 (ordered separately).