KoolTemp® GTS-342 Shipping System, Cold Chain
Supplier: Cold Chain Technologies
GTS-342-SU
89044-500EA 2254.78 USD
89044-500
KoolTemp® GTS-342 Shipping System, Cold Chain
Mailers and Shippers
Insulated container provides universal protection for long-term domestic and international shipment of temperature-sensitive pharmaceuticals, biotech and healthcare products, and medical devices
Prequalified to maintain temperatures of 2 to 8˚C (36 to 46˚F) for 120 hours year-round and in extreme weather conditions. Simple packout process allows quick turnaround of shipments. Constructed from recyclable polyurethane.
Certifications: Pre-qualified in an ISTA-certified and cGMP-compliant lab. Modular components are EPA-compliant, recyclable, replaceable, and reusable under proper conditions.