Pharmaceutical Waste Collector, BD Medical
Supplier: Avantor
89042-024CS 372.97 USD
89042-024
Pharmaceutical Waste Collector, BD Medical
Bins Waste Bins
Dual opening hinge top with 18 VAT perimeter locks allow either side of the collector to be opened depending upon department needs
A proprietary custom-fit process is used to form a foam gasket around the inside of the hinged top, minimizing leaks if the collector is accidentally knocked over