VWR® Air-Cooled Recirculator
Supplier: VWR International
3370P9V11B
89041-314EA 2358.64 USD
89041-314
VWR® Air-Cooled Recirculator
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Economical liquid-to-air cooling unit is ideal for applications where the cooling fluid temperature is higher than the ambient temperature and control is not required.
  • Handles fluid temperatures from ambient to 77°C (170°F)
  • Used in a variety of functions
Commonly used to cool bioreactors, CCD cameras, condensers, ICPs, densitometers, electron microscopes, FTIR, lasers, small injection molds, small Peltier devices, and spectrophotometers. Unit features positive displacement pump, reusable inline fluid filter, built-in low liquid indicator, and integral 0–150psi pressure gauge.
Certifications: cTUVus compliant.