Food BF Latex Fingercots, QRP
Supplier: PIP
BFXL
89041-050CS 1346.75 USD
89041-050
Food BF Latex Fingercots, QRP
Finger Cots Latex Finger Cots
The QRP BF finger cots are manufactured from a proven 100% natural latex formula blended with USDA-compliant blue coloring to eliminate blue rub-off and residue.
  • 100% natural latex, USDA compliant blue coloring
  • For food and poultry handling
  • Skin thin
The BM finger cots are manufactured from a proven 100% natural latex formula blended with USDA-compliant blue coloring to eliminate blue rub-off and residue. BF Series cots are 8 mil thick and offer excellent tactile sensitivity coupled with durability. In addition to protection from skin salts, oils, and other particulates, the high-visibility color ensures easy detection should the cots become dislodged from the handler. Manufactured in FDA-registered facilities utilizing an FDA-registered 510(k) formulation. Meets US Government 21-CFR specificaitons; acceptable for use in food and pharmaceutical applications.
Certifications: Meets US Government 21-CFR specifications.
Caution: This product contains natural rubber latex which may cause allergic reactions.