EP Preserved™ Vials with Preservatives, Thermo Scientific
Supplier: Thermo Scientific
PP140-40CEPPTTW
89039-948CS 788.43 USD
89039-948
EP Preserved™ Vials with Preservatives, Thermo Scientific
Vials
Level 1 glass vials for use in testing for Volatile Organic Compounds (VOC).
Vials are tare-weighed and have pre-applied labels. Propylene caps are open-top with 0.125 PTFE/silicone septa. Capacity: 40 ml. Height: 9.5 cm (3³/₄"). Ø: 2.9 cm (1¹/₈"). Cap Size: 24-414.
Certifications: Vials comply with EPA SW846 Method 5035 requirements.