VWR® Visual Alarm Timer
Supplier: VWR International
93678-89
89038-756EA 28.98 USD
89038-756
VWR® Visual Alarm Timer
Timers Digital Timers
Timer counts down from 9999 minutes in one-minute increments and has a countdown resolution of one second for times under 99 minutes.
  • Bright LED display
  • Multitude of timing features
  • Audible or visual alarms
With the flip of a switch, select either an audible alarm or bright flashing LED visual alarm. Ideal for repetitive tests: automatic memory resets the display to the last programmed time. 'Thru-zero' feature allows counting down, alarming, then counting up. Digits are 1 cm (³/₈") high and readable from 3 m (10').
Certifications: To assure accuracy, an individually serial-numbered Traceable? certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
Ordering information: Supplied with two AAA batteries, integrated magnet, stand, and clip.