ICON® 20 hCG Pregnancy Test, Beckman Coulter®
Supplier: Hemocue
395097A
89038-224CS 1013.43 USD
89038-224
ICON® 20 hCG Pregnancy Test, Beckman Coulter®
Clinical Diagnostic Supplies Clinical Reagents and Kits
Rapid test for detecting hCG qualitatively.
  • Qualitative
  • Sensitive (Serum 10 mIU/mL, Urine 20 mIU/mL )
  • Solid-phase chromatography
  • Detect hCG
Uses a solid-phase chromatographic immunoassay technology to selectively detect elevated levels of hCG in serum or urine. Simple, one-step process for greater reliability and higher sensitivity. Built-in positive and negative controls.
Certifications: Urine is CLIA waived; serum is moderately complex.
Ordering information: Kit contents include 25 test devices and 25 disposable droppers.