VWR® Annealing Cup, Porcelain
Supplier: VWR International
89038-212
89038-212CS 426.04 USD
89038-212
VWR® Annealing Cup, Porcelain
Crucibles
Chemical-porcelain annealing cup is glazed inside and out, except for outside bottom surface and rim.
  • Conical form
  • Glazed interior and exterior except for bottom and rim
  • Chemical-porcelain construction
Smooth interior surfaces in a conical form.