VWR® Long Spatula, Porcelain
Supplier: VWR International
89038-200
89038-200CS 903.99 USD
89038-200
VWR® Long Spatula, Porcelain
Spatulas
Chemical-porcelain tool features a spatula on one end and knob on the other.
  • High chemical resistance
  • Chemical-porcelain construction
  • Glazed for added protection