VWR® Large Desiccator Plate
Supplier: VWR International
89038-054
89038-054CS 256.15 USD
89038-054
VWR® Large Desiccator Plate
Desiccators Desiccators, Plates
Chemical-porcelain plate is glazed on one side and contains eight 30 mm holes.
  • Provided without feet
  • Autoclavable
  • Plate diameter: 230mm