VWR® Rose Crucible, Porcelain
Supplier: VWR International
89038-052
89038-052CS 274.79 USD
89038-052
VWR® Rose Crucible, Porcelain
Crucibles
Unglazed rose crucible without perforated cover or tube.
  • Chemical porcelain construction
  • For use in high-temperature applications
  • Unglazed
  • No perforated cover or tube
For use in temperatures up to 1150°C (2102°F).