VWR® Bitumen Crucible, Porcelain
Supplier: VWR International
89038-036
89038-036CS 96.39 USD
89038-036
VWR® Bitumen Crucible, Porcelain
Crucibles
Chemical-porcelain bitumen crucible features a perforated bottom and large filtering surface.
  • Glazed inside and out
  • Used with 30 to 33 mm diameter filter paper
Crucible is glazed inside and out, except for outside bottom surface and rim. For use with 30–33mm diameter filter paper.