VWR® Midi 10 Electrophoresis System
Supplier: VWR International
E1010-10-VWR
89032-292EA 1086.46 USD
89032-292
VWR® Midi 10 Electrophoresis System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Ideal for processing large numbers of samples in short to medium runs.
  • Durable rubber casting gates
  • Two-piece comb design
  • Small benchtop footprint
  • Durable for rigorous everyday use
System features a small footprint and is built to withstand rigors of everyday use
Durable rubber casting gates permit easy, tape-free casting, while slots in the sides of the trays allow for easy comb placement. Two-piece comb design provides for vertical adjustment, giving control over well depth. Platinum electrode cassettes remove easily for cleaning or replacement.
The "EasyLift" gel box lid is easily removed using the side tabs and pressure pads. Domed lid prevents condensation from dripping onto gel and is surrounded by a drip ring to help recover the condensate and maintain buffer concentration.
Gel box and tray are molded with thick acrylic for leakproof performance. Tray is UV transparent for easy visualization of samples. System accepts 10x10 cm (315/16x315/16") trays. Corrosion-resistant platinum electrodes are held in protective cassettes for safe cleaning and easy replacement.
Ordering information: Supplied with one 10x10 cm (315/16x315/16") gel tray; two 10-tooth, 1.5mm thick combs; casting dams; and well visualization strips.