QBC® STAR Centrifuge Tubes, Drucker Diagnostics
Supplier: Drucker
429625
89031-554PK 1257.17 USD
89031-554
QBC® STAR Centrifuge Tubes, Drucker Diagnostics
Tubes Centrifuge Tubes
QBC® STAR Centrifuge Tubes are designed for use with the QBC® STAR Centrifugal Hematology Analyzer, and hold a 70 µl sample of capillary or venous blood.
Tubes feature pre-inserted ventilated stoppers, and floats contained in the cap.