VWR® Critical Swab® Cone-Shaped Cotton Double Tipped Swab, Paper Handle
Supplier: VWR International
89031-288
89031-288CS 162.82 USD
89031-288
VWR® Critical Swab® Cone-Shaped Cotton Double Tipped Swab, Paper Handle
Swabs Swabs for Sampling
Swab features a paper stem and two cone-shaped tips composed of lintless, highly absorbent cotton.
  • Antistatic packing
  • Lintless, highly absorbent cotton
Packaged in an antistatic leaf pack.
Certifications: CE marked.
Ordering information: Packaged as 50 leaves of 25 swabs per leave.