VWR® Critical Swab® Swab, Large Rectangular Foam Head
Supplier: VWR International
89031-286
89031-286CS 517.33 USD
89031-286
VWR® Critical Swab® Swab, Large Rectangular Foam Head
Swabs Swabs for Sampling
Disposable rectangular swab features 100% polyurethane foam with 100ppi and a sturdy polypropylene handle.
  • Polyurethane foam head
  • Sturdy polypropylene handle
  • Disposable
Thermally bonded foam head reduces contamination.