VWR® Critical Swab® Elongated Foam Tipped Swab, PP Handle
Supplier: VWR International
89031-284
89031-284CS 546.68 USD
89031-284
VWR® Critical Swab® Elongated Foam Tipped Swab, PP Handle
Swabs Swabs for Sampling
Swab features reticulated foam and a blue glass-filled polypropylene shaft for extra rigidity.
  • Polypropylene shaft
  • Reticulated foam
  • Extra rigid
Made in the USA.
For use in critical environments. Handle end pointed for dual use.
Certifications: CE marked.
Packaging: Packaged as 20 bags of 50 swabs per bag.