VWR® Critical Swab® Swab, Foam-Over-Cotton Head
Supplier: VWR International
89031-280
89031-280CS 393.84 USD
89031-280
VWR® Critical Swab® Swab, Foam-Over-Cotton Head
Swabs Swabs for Sampling
Disposable foam-over-cotton tip swab features 100% polyurethane foam with 100 ppi and a wood shaft.
  • Polyurethane foam head
  • Wood shaft