VWR® Critical Swab® Swab, Paddle-Shaped Foam Head
Supplier: VWR International
1876-PFVWR
89031-276CS 409.91 USD
89031-276
VWR® Critical Swab® Swab, Paddle-Shaped Foam Head
Swabs Swabs for Sampling
Disposable, flexible paddle swab features 100% polyurethane foam with 100ppi and blue polypropylene shaft.
  • Polyurethane foam head
  • Blue polypropylene shaft
  • Flexible and disposable