LabX Direct Software, Mettler-Toledo
Supplier: Mettler Toledo
51302876
89031-266EA 680 USD
89031-266 89031-262 11278-504
LabX Direct Software, Mettler-Toledo
Software Laboratory Software
LabX Basic Transfer Software supports the laboratory manager on the PC in the office and the laboratory technician on the touchscreen of the instrument.
  • The software is versatile and easy to use
Balance link allows data communication between a wide range of METTLER TOLEDO instruments; analytical instruments, moisture analyzers, balances and terminals, and pH meters.
Fast and Uncomplicated–With the BalancesLink PC software you can transfer measurement results directly into an application program, a freely chosen input window, or wherever else you position the cursor.
For professional use–Program settings can be saved for future use. Instrument specific configuration options can be protected and only accessed by authorized users.