SP Bel-Art ProCulture Cell Lifter, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37756-0010
89030-910CS 364.68 USD
89030-910
SP Bel-Art ProCulture Cell Lifter, Bel-Art Products, a part of SP
Cell Scrapers
Economical and reusable Cell Lifter scrapes cell lysates and removes and harvests tissue cultures from all types of dishes.
  • Angled, chiseled PP blade
  • Chemically resistant
  • Autoclavable at 121 °C (250 °F)
The narrow blade is ideal for use in 35 mm and 60 mm Petri dishes, as well as 6-well and 12-well cell culture clusters.