FlexTech™ Cut Resistant High Performance Polyethylene Fiber Gloves, Wells Lamont
Supplier: Wells Lamont
Y9265XS Y9265S Y9265M Y9265L Y9265XL Y9266L Y9266M Y9265XXL
89030-788PK 252.76 USD
89030-788 89030-790 89030-792 89030-794 89030-796 80092-410 80092-412 80092-414 75836-536
FlexTech™ Cut Resistant High Performance Polyethylene Fiber Gloves, Wells Lamont
Gloves
FlexTech HPPE gloves with palm coating provide cut resistance and grip.
  • High performance polyethylene fibers (HPPE)
  • Gray polyurethane palm coating
  • Perfect for general assembly
  • Washable for a longer wear life
FlexTech high performance polyethylene fibers (HPPE) and gray nylon blend with gray palm coating provides a good balance of comfort, dexterity, and cut resistance for customers who work in general assembly and applications in which cut resistance is required.
Lint-free multifilament HPPE yarn is lightweight yet 15 times stronger than steel and provides resistance to abrasion, moisture, and chemicals. Polyurethane palm coating provides excellent grip and tactility. These washable gloves have a long wear life.
Certifications: ANSI Level 2.
Ordering information: These gloves are sold by the dozen pair.