Amplitude™ Sigma™ Wipes, Contec®
Supplier: Contec
C1-1212 AMSI0001 AMSI0002 AMSI0001
68310-076CS 501.02 USD
68310-076 89030-186 89030-188 76267-488
Amplitude™ Sigma™ Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Sigma™ Wipes are a hydroentangled blend of 55% cellulose and 45% polyester. They are highly sorbent, offering excellent durability and good cleanliness.
  • Recommended for ISO Class 5 to 8 environments
  • Hydroentangled fabric provides low levels of particles and extractables
  • Excellent for wiping and absorbing spills
  • Strong and durable both wet and dry
  • Cost-effective general-purpose wipe
Amplitude™ Sigma™ wipes are strong and durable wet or dry, making them excellent for use in general cleaning and spill pickup.