SP Bel-Art Techni-Dome® Vacuum Desiccator, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42029-0000
89029-710EA 603.38 USD
89029-710
SP Bel-Art Techni-Dome® Vacuum Desiccator, Bel-Art Products, a part of SP
Desiccators Desiccators, Plastic
This domed desiccator provides a roomy interior for large, moisture-sensitive materials as well as portability and ease of use.
  • For use with moisture-sensitive materials
  • Clear polycarbonate construction
  • Unobsctructed view of contents
  • Resistant against chemicals and thermal stress
Tapered stopcock accepts ¼" to ⅜” I.D tubing and has an easy turning PTFE valve which allows for the gentle entrance of air and minimal disturbance to the inside chamber and its contents.
Used to degas liquids, porous solids, or silicon molds to eliminate air bubbles, it has a spacious 65 liter (4000 cu. in.) internal volume that can hold items up to 45 cm (18”) tall or wide, including racks, trays and instruments. It holds a full vacuum (29” Hg) at room temperature for 24 hours.
Laboratory grade polycarbonate provides a clear, unobstructed view of contents and is resistant to most common chemicals and thermal stress. Side handles provide easy portability and can be used to affix a lock or tamper-evident seals. The unit includes two removable polypropylene shelves; 36.5 cm (14.5”) and 22.5 cm (9”). U.S. Patents: 7,594,342 and D556,895 S.
Ordering information: Gas ports can be closed to use desiccator with standard desiccant cartridges; contact your Avantor representative for more information.